Key Documents
Z280216
Tygon® formula R-3603 laboratory tubing
I.D. × O.D. 3/4 in. × 1 1/2 in.
About This Item
UNSPSC Code:
41100000
packaging
pkg of 10 feet
manufacturer/tradename
Tygon
I.D. × O.D.
3/4 in. × 1 1/2 in.
wall size
3/8 in.
General description
Clear, general-purpose laboratory tubing. Ideal for peristaltic pumps, autoclavable to 248°F for 30 min. Temp. range: -49 to 158°F. Heavy walled Tygon is suitable for vacuum use.
  • Resists most inorganic chemicals
  • Resistant to cracking and aging
  • Low vapor pressure
  • Lasts longer than rubber tubing
Application
suitable for vacuum
Legal Information
Tygon is a registered trademark of Saint-Gobain Performance Plastics Corp.
Choose from one of the most recent versions:
Certificates of Analysis (COA)