Key Documents
Z276839
Screw-cap cuvettes
glass, Open-top cap, spectral range 334-2,500 nm
Synonym(s):
cuvette quartz, quartz cell, quartz cuvette
About This Item
UNSPSC Code:
23151817
material
glass
parameter
334-2,500 nm spectral range
H × W × D
45 mm × 12.5 mm × 12.5 mm
chamber volume
3,500 μL
pathlength
10 mm
General description
  • Solid and open-top cap styles
  • Protect air-sensitive samples
  • Prevent evaporation and contamination
  • Avoid potential spills and exposure to chemicals
Available as a solid-top screw-cap with silicon rubber liner and an open-top screw-cap with silicon rubber septum. Has 2 clear windows and 2 frosted windows. Standard 3.5 mL rectangular cells. Light path: 10 mm. Fits all standard spectrophotometers.
Choose from one of the most recent versions:
Certificates of Analysis (COA)