Key Documents
Z276618
Sigma-Aldrich
Gas cylinder hand truck
no. of cylinders: 2
About This Item
UNSPSC Code:
41100000
NACRES:
NB.51
H
1067 mm
42 in.
weight
65 lb
General description
These collapsible trucks make cylinder handling safe and convenient. Adjustable, spring-locked carriages permit cylinders to be carried at a comfortable angle (45°). Trucks have painted tubular steel frames, cylinder restraining straps, and pneumatic wheels/casters.
Choose from one of the most recent versions:
Certificates of Analysis (COA)