Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z274003

Sigma-Aldrich

FRIO-Temp® bottle thermometer

freezer, temperature -30-1 °C

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41112303

usage

freezer

feature

subdivision 0.5 °C

manufacturer/tradename

HB Instruments 20670

temperature measuring range

-30-1 °C

L

115 mm

Looking for similar products? Visit Product Comparison Guide

General description

NIST traceable Enviro-Safe thermometer with PTFE-coating in an unbreakable plastic bottle, containing clear, Enviro-Safe liquid. The enclosed thermometer accurately records temperature when equipment door is open. Bottles have both Velcro and magnetic attachments for easy installation.

Other Notes

Spirit-filled

Legal Information

FRIO-Temp is a registered trademark of SP Scienceware, a division of SP

related product

Product No.
Description
Pricing

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

152.6 °F

flash_point_c

67 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service