Key Documents
Z273953
Enviro-Safe® thermometer
temperature -10-150 °C, immersion level, total
About This Item
UNSPSC Code:
41112303
NACRES:
NB.79
feature
immersion level total
manufacturer/tradename
HB Instruments 20594
temperature measuring range
-10-150 °C
L
200 mm
General description
Enviro-Safe thermometers are serialized, calibrated with NIST (NBS) traceable standards, and include a Statement of Accuracy.
White glass with easy-to-read, non-hazardous, biodegradable, green liquid fill. 6.5 mm O.D. guaranteed accuracy: ±1 scale division below 105°C (221°F), ±1.5 scale divisions above 105°C (221°F), and ±2 scale divisions above 200°C (392 °F).
Legal Information
Enviro-Safe is a registered trademark of H-B Instrument Co.
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)