Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

Z273945

Sigma-Aldrich

Enviro-Safe® thermometer

greener alternative

temperature -10-150 °C, immersion level, 50 mm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41112303
NACRES:
NB.79

feature

immersion level 50 mm

manufacturer/tradename

HB Instruments 20593

greener alternative product characteristics

Designing Safer Chemicals
Learn more about the Principles of Green Chemistry.

temperature measuring range

-10-150 °C

L

200 mm

greener alternative category

Looking for similar products? Visit Product Comparison Guide

General description

Enviro-Safe thermometers are serialized, calibrated with NIST (NBS) traceable standards, and include a Statement of Accuracy.

White glass with easy-to-read, non-hazardous, biodegradable, green liquid fill. 6.5 mm O.D. guaranteed accuracy: ±1 scale division below 105°C (221°F), ±1.5 scale divisions above 105°C (221°F), and ±2 scale divisions above 200°C (392 °F).
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product is designed as a safer chemicals without mercury.

Legal Information

Enviro-Safe is a registered trademark of H-B Instrument Co.

related product

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service