Z273937
Enviro-Safe® thermometer
temperature -10-110 °C, immersion level, total
All Photos(2)
About This Item
UNSPSC Code:
41112303
NACRES:
NB.79
feature
immersion level total
manufacturer/tradename
HB Instruments 20592
greener alternative product characteristics
Designing Safer Chemicals
Learn more about the Principles of Green Chemistry.
temperature measuring range
-10-110 °C
L
200 mm
General description
Enviro-Safe thermometers are serialized, calibrated with NIST (NBS) traceable standards, and include a Statement of Accuracy.
White glass with easy-to-read, non-hazardous, biodegradable, green liquid fill. 6.5 mm O.D. guaranteed accuracy: ±1 scale division below 105°C (221°F), ±1.5 scale divisions above 105°C (221°F), and ±2 scale divisions above 200°C (392 °F).
White glass with easy-to-read, non-hazardous, biodegradable, green liquid fill. 6.5 mm O.D. guaranteed accuracy: ±1 scale division below 105°C (221°F), ±1.5 scale divisions above 105°C (221°F), and ±2 scale divisions above 200°C (392 °F).
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product is designed as a safer chemicals without mercury.
Legal Information
Enviro-Safe is a registered trademark of H-B Instrument Co.
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)