For Natural Gas. Nickel plated brass burner tube 13 mm diameter with rotatable non-removable air regulator, on stable cast iron base, baked hammertone finish, with serrated gas inlet tube.
Laboratory gas burners for use with natural (methane) or cylinder (propane) gases.
Available in 3 styles:
General-purpose - for general laboratory applications (solid base)
Fast-flame - with flame stabilizer to control flame temperature and length (three spoke base)
Meker - for applications where an extremely hot flame is required (only for bottled gas, perforated metal mesh burner)
Choose from one of the most recent versions:
Certificates of Analysis (COA)