Key Documents
Z270288
Sigma-Aldrich
Bunsen burner
Natural gas: General-purpose
Synonym(s):
bunsen burner gas, bunsen burner lab, bunsen burners
About This Item
UNSPSC Code:
41100000
General description
For Natural Gas. Nickel plated brass burner tube 11 mm diameter with rotatable non-removable air regulator. On stable cast iron base, baked hammertone finish, with serrated gas inlet tube 9 mm outer diameter.
Laboratory gas burners for use with natural (methane) or cylinder (propane) gases.
Available in 3 styles:
  • General-purpose - for general laboratory applications (solid base)
  • Fast-flame - with flame stabilizer to control flame temperature and length (three spoke base)
  • Meker - for applications where an extremely hot flame is required (only for bottled gas, perforated metal mesh burner)
Choose from one of the most recent versions:
Certificates of Analysis (COA)