Key Documents
Z269727
Sigma-Aldrich
Scienceware® Frigimat® dry ice makers
European connectors
About This Item
UNSPSC Code:
41103000
NACRES:
NB.22
material
aluminum ice chamber
manufacturer/tradename
Bel-Art
General description
Produces solid 1 lb cylindrical block 8.5 × 11 cm of dry ice from a cylinder of liquid CO2. One CO2 cylinder makes 8 to 9 blocks of dry ice. Epoxy-coated steel frame, brass valve, aluminum dry ice chamber.
Legal Information
Frigimat is a registered trademark of SP Scienceware, a division of SP
Scienceware is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)