Key Documents
Z269573
Safety pipette fillers
red
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
material
natural rubber
manufacturer/tradename
FLIP-001
color
red
General description
Has two operating valves to fill and dispense and a third removable automatic valve at the top for evacuation and easy inside cleaning. Natural rubber.
Choose from one of the most recent versions:
Certificates of Analysis (COA)