Key Documents
Z269026
Sigma-Aldrich
Scienceware® Safe-Lab glass stopper
joint: ST/NS 29/42
Synonym(s):
flask stopper, glass stoppers, stoppers
About This Item
UNSPSC Code:
41122500
NACRES:
NB.35
joint
joint: ST/NS29/42
manufacturer/tradename
Bel-Art
General description
Lightweight, hollow, borosilicate ground-glass stoppers with PP extracting nut. Flat hex-head for easy gripping. Interchangeable with pennyhead stoppers.
Legal Information
Safe-Lab is a trademark of SP Scienceware, a division of SP
Scienceware is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)