Z268852
Scienceware® Safe-Lab™ vacuum adapter
joint: ST/NS 24/40
All Photos(1)
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
joint
joint: ST/NS24/40
manufacturer/tradename
Bel-Art
General description
PTFE body with Viton® O-ring. Hose barb connection for vacuum work or inlet/outlet of gases.
Legal Information
Safe-Lab is a trademark of SP Scienceware, a division of SP
Scienceware is a registered trademark of SP Scienceware, a division of SP
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)