material
ZnSe
packaging
pkg of (contains 1 window)
technique(s)
IR spectroscopy: suitable
diam. × thickness
32 mm × 3 mm
General description
Cut and polished for demountable liquid and gas cell holders
Choose from one of the most recent versions:
Certificates of Analysis (COA)