Key Documents
Z267368
Hamilton® syringe barrel
for use with Luer tip syringe 1005, volume 5 mL
About This Item
UNSPSC Code:
41122001
NACRES:
NB.03
material
glass
description
1005
feature
for LT syringes (5 mL)
packaging
pkg of 1 ea
manufacturer/tradename
Hamilton 10123
volume
5 mL
compatibility
for use with Luer tip syringe 1005
General description
Hamilton syringe is available in gas-tight type that minimizes the gas exchange in analysis. It gives accurate volume delivery. The plunger is mostly unsurpassed.[1] The tip is threaded in a luer lock syringe allowing the needle to be tightly screwed to the tip.[2]
Legal Information
Hamilton is a registered trademark of Hamilton Co.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Customers Also Viewed
Jeffrey S. Guy
Pharmacology for the Prehospital Professional, 54-54 (2009)
In Vitro Fertilization and Embryo Transfer: A Manual of Basic Techniques
Don P. Wolf
Springer Science & Business Media, 192-192 (2012)