Key Documents
Z266078
Supelco
PTFE multi-plate racks
For use with 10 cm × 10 cm plates
About This Item
UNSPSC Code:
41115711
NACRES:
NC.07
technique(s)
thin layer chromatography (TLC): suitable
General description
For use with Latch-Lid developing chamber. Can accommodate up to six TLC plates.
Legal Information
Latch-Lid is a trademark of General Glassblowing, Inc.
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)