Key Documents
Z266027
Supelco
TLC plate rack
About This Item
UNSPSC Code:
41115711
NACRES:
NC.07
material
stainless steel rack
technique(s)
thin layer chromatography (TLC): suitable
General description
Holds two 20 × 20 cm, 10 × 20 cm, or 5 × 20 cm TLC plates in a standard glass developing chamber.
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)