Key Documents
Z265284
Supelco
TLC plate holder
About This Item
UNSPSC Code:
41115711
NACRES:
NC.07
manufacturer/tradename
Analtech 5002
technique(s)
thin layer chromatography (TLC): suitable
General description
Horizontal and vertical grooves hold 25 TLC plates of any size in an upright position. Lightweight body resistant to usual TLC solvents and chemicals. Great for desiccator storage of plates.
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)