Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z264792

Sigma-Aldrich

Hydrion® Brilliant pH dip stiks

pH range 6.5-13.0

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41113034
NACRES:
NB.79

feature

non-bleeding: no
refill: no

packaging

pkg of 100 ea

manufacturer/tradename

Hydrion

graduations and accuracy measuring range

accuracy: 0.5 pH unit

pH range

6.5-13.0

detection method

colorimetric

Looking for similar products? Visit Product Comparison Guide

General description

Direct reading pH stick with clear, bright color matches over the desired pH range.

Legal Information

Hydrion is a registered trademark of Micro Essential Laboratory, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service