Z261041
Nalgene® polypropylene copolymer separatory funnel
capacity 1,000 mL
All Photos(1)
About This Item
UNSPSC Code:
41121809
NACRES:
NB.43
Recommended Products
capacity
1,000 mL
closure size
53 mm
General description
Break-resistant and nearly transparent. Phase interface visible down to stopcock. PTFE stopcock and Tefzel® screw closure are leakproof.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Tefzel is a registered trademark of E. I. du Pont de Nemours and Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)