Key Documents
Z258164
Sigma-Aldrich
Polyethylene gloves
size M, shoulder length
About This Item
UNSPSC Code:
46181504
size
M
packaging
pack of 100 gloves
availability
not available in EU
thickness
1.0 mil
General description
Talc-free. Thin gauge poly film allows keen touch sensitivity and short-term protection from moisture and chemicals. Ambidextrous, disposable. Cannot be used for cell culture applications.
Choose from one of the most recent versions:
Certificates of Analysis (COA)