Z257818
Enviro-Safe® pocket thermometer
temperature 0-220 °F, With armor
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41112303
feature
subdivision 2 °F
manufacturer/tradename
HB Instruments 20580
greener alternative product characteristics
Designing Safer Chemicals
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
temperature measuring range
0-220 °F
L
145 mm
greener alternative category
, Aligned
Looking for similar products? Visit Product Comparison Guide
General description
Total immersion. With green, non-hazardous, liquid fill. 160 mm L with aluminum case. An armor option is available, providing extra shielding for the thermometer.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product is designed as a safer chemicals without mercury.
Legal Information
Enviro-Safe is a registered trademark of H-B Instrument Co.
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service