Key Documents
Z257583
Sigma-Aldrich
PROHEAT® VARITEMP® heat guns
PH-1200, AC/DC input 120 V AC, 12 A
About This Item
UNSPSC Code:
41102404
AC/DC input
120 V (AC)
material
plastic
feature
CSA approved
availability
not available in EU
parameter
453 L/min flow rate
49-449 °C temp. range
General description
These heat guns offer all the features of the PROHEAT heat guns, plus variable temperature control. Dial in any temperature from 130°F (49°C) to 840°F (449°C). Ideal for confined spaces or for applications where its lightweight construction can prevent operator fatigue.
Legal Information
PROHEAT is a registered trademark of Master Appliance Corp.
VARITEMP is a registered trademark of Master Appliance Corp.
Choose from one of the most recent versions:
Certificates of Analysis (COA)