Key Documents
Z257567
Sigma-Aldrich
PROHEAT® heat gun
PH-1100, AC/DC input 120 V AC, 6 - 12 A
About This Item
UNSPSC Code:
41102404
NACRES:
NB.76
AC/DC input
120 V (AC)
feature
CSA approved
availability
not available in EU
parameter
540 °C temp. range
General description
Compact, lightweight heat guns with dual temperature settings of 430°F and 840°F (221°C and 449°C). Excellent source of flameless heat with air flow at 15 cfm. The PROHEAT heat guns meet most general laboratory applications.
Legal Information
PROHEAT is a registered trademark of Master Appliance Corp.
Choose from one of the most recent versions:
Certificates of Analysis (COA)