Key Documents
Z256048
Gum rubber laboratory tubing
tubing I.D. × O.D. 1 in. × 1 3/4 in.
About This Item
UNSPSC Code:
41100000
description
heavy wall
packaging
pkg of 10 feet
tubing I.D. × O.D.
1 in. × 1 3/4 in.
wall size
3/8 in.
General description
Tubing is red, and can be used for pressure or vacuum work.
Choose from one of the most recent versions:
Certificates of Analysis (COA)