Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z255890

Pyrex® glass fiber wool

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
41121800
NACRES:
NB.43
Product Z255890 is not currently sold in your country. Contact Technical Service

material

borosilicate glass membrane
roll

manufacturer/tradename

SciLabware 3950

diam. × L

2 in. × 22 ft

pore size

8 μm

General description

Made from extremely fine borosilicate glass-fibers with 8 μm diam. pore size.

Strong, soft and pliable; free of heavy metals, with extremely low levels of fluorine and alumina; low alkali content. Suitable for chromatography use, as a filtering medium, or as an "inert sponge" to reclaim small chemical spills in the lab.

Supplied in woven roving.

Packaging

Pkg = 1lb

Legal Information

PYREX is a registered trademark of Corning, Inc.

replaced by

pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Carc. 1B Inhalation

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service