Z253006
Enviro-Safe® thermometer
temperature -20-150 °C, immersion level, total
About This Item
Recommended Products
feature
immersion level total
manufacturer/tradename
HB Instruments 20510
greener alternative product characteristics
Designing Safer Chemicals
Learn more about the Principles of Green Chemistry.
temperature measuring range
-20-150 °C
L
305 mm
General description
White glass with easy-to-read, non-hazardous, biodegradable, green liquid fill. 6.5 mm O.D. guaranteed accuracy: ±1 scale division below 105°C (221°F), ±1.5 scale divisions above 105°C (221°F), and ±2 scale divisions above 200°C (392 °F).
Legal Information
related product
Choose from one of the most recent versions:
Certificates of Analysis (COA)