Key Documents
Z252417
Supelco
Wheaton vial rack
15 mm vial, no. of holes, 48 (4 × 12)
About This Item
UNSPSC Code:
41122001
feature
no. of holes 48 (4 × 12)
packaging
pkg of 5 ea
L × W × H
9.4 cm × 26.6 cm × 2.8 cm
General description
Chemically resistant polypropylene racks have corner posts to ensure stability when stacking empty. Autoclavable at 121°C for 20min. Vial grid is alphanumerically indexed for easy sample identification.
Choose from one of the most recent versions:
Certificates of Analysis (COA)