description
semi-quantitative
specific analyte(s)
zinc
packaging
pack of 100 ea
manufacturer/tradename
Macherey-Nagel
detection method
colorimetric
General description
- Fast; dip and read
- No apparatus needed, only single test stick for analysis
- Highly specific; interferences can be eliminated by precipitation or masking
- Dim.: 6 × 95 × 0.2 mm
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)