Key Documents
Z247715
Coors table-type removable plate Buchner funnel
Perforated plate
About This Item
UNSPSC Code:
41121809
NACRES:
NB.43
material
porcelain
packaging
pack of 1 ea
manufacturer/tradename
Coorstek 60264
L × W × H
19 cm (7 1/2 in.) × 7.6 cm (3 in.) × 12 cm (4 3/4 in.)
weight
0.8 lb
General description
Porcelain. Removable perforated plate allows thorough cleaning. Side-outlet for vacuum. Glazed inside and out except for rim and seat portion of plate and funnel. Order funnel and perforated plate separately.
The Scienceware® Buchner funnel trowel is specifically designed for use with 18, 24 and 36 in. Buchner funnels. Just apply slight downward pressure and ′sweep′ the trowel along the surface of the precipitate to quickly and efficiently smooth fissures appearing during the filtration process.
  • Large surface area smooths fissures faster than traditional narrow spatula method
  • Dual-beveled edges for bi-directional smoothing
  • Curved ends match Buchner funnel sidewalls allowing easy access to entire cake surface
  • FDA-grade high-density polyethylene (HDPE) complies with industry standards and provides maximum chemical-resistance
  • Welded construction with no hollow cavities assures hygienic use and easy cleaning
  • Compatible with industrial dishwasher temperatures and cleaners
Legal Information
Coors is a trademark of Coorstek, Inc.
Scienceware is a registered trademark of SP Scienceware, a division of SP
accessory
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)