material
circles
porcelain
sterility
non-sterile
packaging
pack of 6 ea
manufacturer/tradename
Coorstek 60202
H
45 mm
capacity
250 mL
dish diam.
115 mm
top O.D.
115 mm
General description
Porcelain, with lip. Glazed inside and out, except for rim. Max. use temp.: 1,150 °C; limit of use evacuated: 1,300 °C.
Legal Information
Coors is a trademark of Coorstek, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)