L
H × W × D
14 in. × 5 in. × 6 in.
General description
White acrylic. Clear hinged top and front windows. Ideal for dust-free storage and dispensing of small items such as rubber septa, disposable pipette tips, centrifuge tubes, etc.
Legal Information
SIGMA-ALDRICH is a registered trademark of Merck KGaA, Darmstadt, Germany
Choose from one of the most recent versions:
Certificates of Analysis (COA)