Z245062
Bransonic® ultrasonic cleaner
Branson 5510EMTH, AC/DC input 220 V AC, tank capacity 9.2 L, with mechanical timer and heater
Synonym(s):
ultrasonic bath sonicator, ultrasonic cleaning bath
About This Item
feature
CE compliant
manufacturer/tradename
Branson 5510EMTH
W × D × H
11 1/2 in. × 6 in. × 9 1/2 in.
tank capacity
9.2 L
General description
Order accessory trays separately.
- Standard models have a single control to activate ultrasonics and set a time limit
- Touch control on digital readout models permits setting and monitoring cleaning cycles up to 99 min, and displays solution temperature in °C
- A degas function is available on these models for immediate use at maximum efficiency
- All units have dependable solid-state circuitry
- Heater temperature 80°C
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)