Key Documents
Z238155
Sigma-Aldrich
Aldrich® LB-cylinder holder
Bench/wall mount, Holds 1 x cylinder
About This Item
UNSPSC Code:
41100000
NACRES:
NB.51
material
PVC
feature
Holds 1 x cylinder
manufacturer/tradename
Sigma-Aldrich Z238155
W × H × D
5 in. × 7 3/4 in. × 4 in.
General description
Hold up to 2 in. diameter cylinders. Bench/wall mountable units have four countersunk screw holes for attachment.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)