Key Documents
Z236217
Supelco
Fine wire kit
10 each of the following sizes: 0.15mm, 0.25mm, and 0.50mm
About This Item
UNSPSC Code:
41122001
availability
not available in EU
General description
The fine wire kit cleans syringes, needles, pens, etc. General-purpose cleaner for fine openings. Includes pin vise and 30 SS wires, 10 each of the following sizes: 0.15mm, 0.25mm, and 0.50mm.
Choose from one of the most recent versions:
Certificates of Analysis (COA)