Key Documents
Z236179
Supelco
FID Cleaning Kit
About This Item
UNSPSC Code:
12352200
availability
not available in EU
General description
Includes three spiral jet reamers, a brass and a stainless steel minibrush, and a dual-ended handle the reamers and brushes.
Choose from one of the most recent versions:
Certificates of Analysis (COA)