Key Documents
Z236144
Supelco
Injection Port Cleaning Kit (with Large Diameter Brushes)
About This Item
UNSPSC Code:
23151817
NACRES:
SB.54
availability
not available in EU
General description
Includes three stainless steel brushes (5 mm, 1/4 in., and 3/8 in. diameters) and one scraper for removing septa residue.
Choose from one of the most recent versions:
Certificates of Analysis (COA)