Key Documents
Z236136
Supelco
Septum Pick
About This Item
UNSPSC Code:
23151817
NACRES:
SB.54
General description
The septum pick is useful for removing small pieces of septa from injection ports, and graphite ferrules from column nuts.
Choose from one of the most recent versions:
Certificates of Analysis (COA)