Z234214
Sigma-Aldrich® TLC Plates
aluminum oxide matrix
About This Item
Recommended Products
material
aluminum oxide matrix
polyester support
feature
binder Organic
fluorescent indicator
packaging
pkg of 25 ea
manufacturer/tradename
Sigma-Aldrich
technique(s)
thin layer chromatography (TLC): suitable
L × W
20 cm × 20 cm
layer thickness
200 μm
particle size
<60 μm
Looking for similar products? Visit Product Comparison Guide
General description
- Silica gel - for weakly to strongly polar separations, pore size 60 Å
- High purity silica gel - acid washed for separation of aflatoxins
- Cellulose - for partition chromatography, (SigmaCell Type 100 Cellulose)
- Cellulose PEI - for separation of weak anions (amino acids, peptides) PEI plates may turn slightly yellow, affecting separation. Pre-develop plates in distilled water to minimize yellow color.
- Chiral silica - for separation of optically active isomers by ligand exchange, pore size 60Å
Other Notes
105581
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service