Key Documents
Z234117
Sigma-Aldrich
Kilo-Lab® cylinder
Diptubes, for cylinder 90 L
About This Item
UNSPSC Code:
41100000
NACRES:
NB.43
General description
For storing and dispensing laboratory-scale to production quantities of air-sensitive liquids.
Technical Information Bulletin No. AL-149, with instructions for use of the system and a list of fittings and accessories, is included with each cylinder. Meets DOT 4BA-240, rated 240 psi.
  • Cylinder does not come with valves won′t have valves
  • Cylinder is not TC stamped as required in Canada
  • The 90 L cylinder is pictured
SS, one end 1/4 in. NPTM; other end angle-cut.
Legal Information
Kilo-Lab is a registered trademark of Merck KGaA, Darmstadt, Germany
Choose from one of the most recent versions:
Certificates of Analysis (COA)