Key Documents
Z231037
Sigma-Aldrich
Aldrich® Library of 13C and 1H FT-NMR Spectra
C. Pouchert and J. Behnke, Aldrich Chemical Co., 1993, 4300 pp., hard cover
About This Item
ISBN-10:
0-94163334-9
ISBN-13:
978-0-94163334-5
UNSPSC Code:
55101509
publication info
C. Pouchert and J. Behnke, Aldrich Chemical Co., 1993, 4300 pp., hard cover
General description
This three volume set of 12,000 high-resolution 75MHz 13C and 300MHz 1H FT-NMR spectra is arranged according to functionality.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)