Key Documents
Z225355
Sigma-Aldrich
Plier-type decapper
O.D. 8 mm
About This Item
UNSPSC Code:
41122001
NACRES:
NB.11
O.D.
8 mm
General description
Removes aluminum seals from serum bottles and vials.
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)