Attain extremely precise, reproducible flow control of both liquids and gases. Valves have a precision-bore Pyrex® glass capillary tube, PTFE rod and stem, and Viton® O-ring. Achieve semilogarithmic flow control in the first 97% of travel with micrometer valve; 70% with metering valve. Micrometer valve includes Viton O-rings for 10-6 mm Hg vacuums and PTFE O-rings for 10-2 mm Hg vacuums. Simple-design metering valve works with high vacuums. Quick-opening metering valve allows fast opening and closing with a small degree of flow regulation. All valves withstand maximum pressures of 30 psi and maximum temperatures of 250 °F (121°C).
Flow control obtained over 70% of the travel. Suitable for high vacuum (10-6 mm Hg) work.
Legal Information
PYREX is a registered trademark of Corning, Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)