Key Documents
Z224480
Sigma-Aldrich
PTFE coupling adapter
NPTF 1/4 in.
About This Item
UNSPSC Code:
20121904
NACRES:
NB.51
manufacturer/tradename
Gilmont
NPTF
1/4 in.
General description
All PTFE adapters attach to threaded PP bushings on flowmeter to provide NPTF ports for pipe-to-tubing adapters below. Internal PTFE O-ring ensures that fluids will not contact PP bushings. Two coupling adapters required per flowmeter. Order PFA pipe-to-tubing adapters as needed below.
For micro, 0, 1, 2, 3, 10, 11, 12, and 13 flowmeters
Choose from one of the most recent versions:
Certificates of Analysis (COA)