Key Documents
Z224278
Sigma-Aldrich
Nylon tubing clamp
diam. 1 3/32-1 3/16 in.
About This Item
UNSPSC Code:
41100000
diam.
1 3/32-1 3/16 in.
General description
Non-corroding, lightweight clamps secure tubing to transfer needles, fittings, etc. Press clamp jaws together with pliers to secure.
Choose from one of the most recent versions:
Certificates of Analysis (COA)