diam.
3/4- 7/8 in.
General description
Non-corroding, lightweight clamps secure tubing to transfer needles, fittings, etc. Press clamp jaws together with pliers to secure.
Choose from one of the most recent versions:
Certificates of Analysis (COA)