Z224189
Chem-FLEX™ tubing clamp
for tubing o.d., 0.38 ‑ 0.47 in.
All Photos(1)
About This Item
UNSPSC Code:
41105108
NACRES:
NB.35
description
for tubing CHEM-FLEX 187
feature
for tubing o.d. 0.38 ‑ 0.47 in.
General description
Noncorroding, lightweight nylon clamps secure CHEM-FLEX tubing to transfer needles and fittings.
Legal Information
Chem-Flex is a trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)