Key Documents
Z223581
Sigma-Aldrich
Aldrich® septum-inlet adapter, female
Plain, joint: ST/NS 24/40
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
joint
joint: ST/NS24/40
General description
Septum-inlets require septum Z565695 or Z565679 except where noted.
Linkage
See the Threaded Ground Glass Tutorial to see how threaded joints work.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)