Key Documents
Z223174
Sigma-Aldrich
Sound abating enclosure
For 500 and 750W models
About This Item
UNSPSC Code:
41103511
NACRES:
NB.74
manufacturer/tradename
Sonics & Materials 630-0427
General description
Greatly reduces high-pitched noise produced by harmonics emanating from vessel walls and fluid surface during processing. Probe assembly is introduced through opening at top. Two access ports allow coolant or sample flow through the processing vessel while the door is closed. Waterproof, acoustically insulated interior and transparent access door.
Choose from one of the most recent versions:
Certificates of Analysis (COA)