Key Documents
Z223069
Extender for 500 and 750W ultrasonic processor
for horn 3/4 in.
About This Item
UNSPSC Code:
41101500
NACRES:
NB.74
manufacturer/tradename
Sonics & Materials 630-0409
General description
Screw into the threaded end of horn, extending the horn length by 5 in., for processing samples up to 7 in. down into narrow vessels. Often used with solvents or low surface tension liquids such as methylene chloride.
Choose from one of the most recent versions:
Certificates of Analysis (COA)